Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST3GAL1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105446 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-446 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-446 Supplier Abnova Corporation Supplier No. H00006482A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant ST3GAL1.

The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein is normally found in the Golgi but can be proteolytically processed to a soluble form. Correct glycosylation of the encoded protein may be critical to its sialyltransferase activity. This protein, which is a member of glycosyltransferase family 29, can use the same acceptor substrates as does sialyltransferase 4B. Two transcript variants encoding the same protein have been found for this gene. Other transcript variants may exist, but have not been fully characterized yet. [provided by RefSeq

Sequence: SHTMVATTWFPKQMVLELSENLKRLIKHRPCTCTHCIGQRKLSAWFDERFNQTMQPLLTAQNALLEDDTYRWWLRLQREKKPNNLNDTIKELFRVVPGNV

Specifications

Antigen ST3GAL1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ST3GAL1.
Formulation 50% glycerol
Gene ST3GAL1
Gene Accession No. NM_003033
Gene Alias DKFZp666E036/DKFZp779K2051/FLJ36548/Gal-NAc6S/MGC9183/SIAT4A/SIATFL/ST3GalA/ST3GalA.1/ST3GalIA/ST3GalIA,1/ST3O
Gene Symbols ST3GAL1
Host Species Mouse
Immunogen ST3GAL1 (NP_003024, 29 a.a. ∽ 128 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6482
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.