Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT1, Mouse, Clone: PSM1, Abnova™

Catalog No. 89135486 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-486 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-486 Supplier Abnova Corporation Supplier No. MAB3641
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic phosphopeptide of STAT1.

Specifications

Antigen STAT1
Applications Immunoprecipitation
Classification Monoclonal
Clone PSM1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic phosphopeptide of STAT1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Synthetic phosphopeptide corresponding to amino acids 721-733 residues surrounding S727 of STAT1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes transcriptional factor STAT1 (91 KDa) activated byphosphorylation at Ser727.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.