Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stat2, Mouse, Clone: 19G8.H2.H6, Abnova™

Catalog No. 89113371 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-371 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-371 Supplier Abnova Corporation Supplier No. MAB8168
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of Stat2.

Specifications

Antigen Stat2
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 19G8.H2.H6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of Stat2.
Dilution ELISA (1:5000-1:10000) Western Blot (1:1000) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene Stat2
Gene Alias 1600010G07Rik/AW496480
Gene Symbols Stat2
Host Species Mouse
Immunogen A synthetic peptide corresponding to C-terminus of mouse Stat2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20847
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.