Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stat5a, Mouse, Clone: 5F6.F1, Abnova™

Catalog No. 89110818 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-818 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-818 Supplier Abnova Corporation Supplier No. MAB2097
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic phosphopeptide of Stat5a.

Specifications

Antigen Stat5a
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 5F6.F1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic phosphopeptide of Stat5a.
Dilution ELISA (1:20000) Western Blot (1:500-1:2000) Immunohistochemistry (20-40 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene Stat5a
Gene Alias AA959963/STAT5
Gene Symbols Stat5a
Host Species Mouse
Immunogen Synthetic phosphopeptide corresponding to residues surrounding Y694 of mouse Stat5a.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20850
Test Specificity This antibody is specific to mouse Stat5alpha protein phosphorylated at Y694.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.