Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STK38, Mouse, Clone: 2G8-1F3, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant STK38.
Supplier: Abnova Corporation H00011329M01A
Description
Sequence: MAMTGSTPCSSMSNHTKERVTMTKVTLENFSSNLIAQHEEREMRQKKLEKVMEEEGLKDEEKRLRRSAHARKETEFLRLKRTRLGLEDFESLKVIGRGAFGEVRLVQKKDTGHVYAMKILRKADMLEKEQVGHIRAERDILVEADSLWVVKMFYSFQDKLNLYLIMEFLPGGDMMTLLMKKDTLTEEETQFYIAETVLAIDSIHQLGFIHRDIKPDNLLLDSKGHVKLSDFGLCTGLKKAHRTEFYRNLNHSLPSDFTFQNMNSKRKAETWKRNRRQLAFSTVGTPDYIAPEVFMQTGYNKLCDWWSLGVIMYEMLIGYPPFCSETPQETYKKVMNWKETLTFPPEVPISEKAKDLILRFCCEWEHRIGAPGVEEIKSNSFFEGVDWEHIRERPAAISIEIKSIDDTSNFDEFPESDILKPTVAISNHPETDYKNKDWVFINYTYKRFEGLTARGAIPSYMKAAKSpecifications
STK38 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
BC012085 | |
STK38 | |
STK38 (AAH12085,1 a.a. ∽ 465 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
11329 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
2G8-1F3 | |
Mouse monoclonal antibody raised against a full-length recombinant STK38. | |
STK38 | |
NDR/NDR1 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction