Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STOM, Mouse, Clone: 13, Abnova™

Catalog No. 89113932 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-932 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-932 Supplier Abnova Corporation Supplier No. MAB9974
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant STOM.

Specifications

Antigen STOM
Applications ELISA, Western Blot
Classification Monoclonal
Clone 13
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant STOM.
Dilution Western blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300)
Gene STOM
Gene Alias BND7/EPB7/EPB72
Gene Symbols STOM
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human STOM.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2040
Test Specificity It can expression in HEK293 whole cell lysate.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.