Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STRO-1, Mouse, Clone: STRO-1, Abnova™

Catalog No. 89134809 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-809 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-809 Supplier Abnova Corporation Supplier No. MAB10247
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against of STRO-1.

Specifications

Antigen STRO-1
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone STRO-1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against of STRO-1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In TBS, pH 8.0 (0.09% sodium azide)
Host Species Mouse
Immunogen Human CD34 positive bone marrow cells.
Purification Method Precipitation methods and ion exchange chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The mouse monoclonal antibody STRO-1 recognizes the cell surface antigen STRO-1 expressed by bone marrow mesenchymal stromal cells and nucleated erythroid precursors, but not by committed hematopoietic progenitors.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.