Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SULF1, Mouse, Clone: 1A4, Abnova™

Catalog No. 89106570 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-570 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-570 Supplier Abnova Corporation Supplier No. H00023213M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SULF1.

Heparan sulfate proteoglycans (HSPGs) act as coreceptors for numerous heparin-binding growth factors and cytokines and are involved in cell signaling. Heparan sulfate 6-O-endosulfatases, such as SULF1, selectively remove 6-O-sulfate groups from heparan sulfate. This activity modulates the effects of heparan sulfate by altering binding sites for signaling molecules (Dai et al., 2005 [PubMed 16192265]).[supplied by OMIM

Sequence: RTVNETHNFLFCEFATGFLEYFDMNTDPYQLTNTVHTVERGILNQLHVQLMELRSCQGYKQCNPRPKNLDVGNKDGGSYDLHRGQLWDGWEG

Specifications

Antigen SULF1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1A4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SULF1.
Formulation ascites with no preservative
Gene SULF1
Gene Accession No. NM_015170
Gene Alias FLJ30905/FLJ38022/FLJ41750/HSULF-1/KIAA1077/SULF-1
Gene Symbols SULF1
Host Species Mouse
Immunogen SULF1 (NP_055985.1,780 a.a. ∽ 871 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23213
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.