Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SURF6, Mouse, Clone: S-208, Abnova™

Catalog No. 89108453 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-453 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-453 Supplier Abnova Corporation Supplier No. MAB10250

Mouse monoclonal antibody raised against recombinant SURF6

This gene is located in the surfeit gene cluster, a group of very tightly linked genes that do not share sequence similarity. The gene demonstrates features of a housekeeping gene, being ubiquitously expressed, and the encoded protein has been localized to the nucleolus. The protein includes motifs found in both the mouse and fish orthologs, which suggests a putative function as a nucleolar-matrix protein with nucleic acid-binding properties, based on characteristics determined in mouse. [provided by RefSeq

Specifications

Antigen SURF6
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone S-208
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant SURF6
Dilution ELISA (1:30-1:3000) immunoprecipitation (1 ug per 1 mg of lysate) Western Blot (1:100-1:5000) Immunohistochemistry (Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:500) Immunofluorescence (1:150-1:500) The opti
Formulation In PBS, pH 7.4 (0.05% sodium azide)
Gene SURF6
Gene Alias FLJ30322
Gene Symbols SURF6
Host Species Mouse
Immunogen Recombinant GST fusion protein corresponding to human SURF6.
Purification Method IgG purification
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6838
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.