Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SV40 tag, Mouse, Clone: 8f7s9, Abnova™

Catalog No. 89136602 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-602 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-602 Supplier Abnova Corporation Supplier No. MAB8761
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native SV40 tag.

Specifications

Antigen SV40 tag
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 8f7s9
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native SV40 tag.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.02% sodium azide)
Host Species Mouse
Immunogen Native purified C-terminus amino acids of SV40 tag.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with native and denatured large T antigen. It does not react with small T antigen of SV40 or large T antigen of BKV and human papovavirus.
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.