Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYK, Mouse, Clone: 4D10.1, Abnova™

Catalog No. 89110641 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-641 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-641 Supplier Abnova Corporation Supplier No. MAB1806

Mouse monoclonal antibody raised against synthetic peptide of SYK.

Specifications

Antigen SYK
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4D10.1
Conjugate Unconjugated
Description spleen tyrosine kinase
Formulation In PBS (0.08% sodium azide)
Gene SYK
Gene Alias DKFZp313N1010, FLJ25043, FLJ37489
Gene Symbols SYK
Host Species Mouse
Immunogen A synthetic peptide corresponding to cytoplasmic domain of human SYK.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6850
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.