Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYK, Mouse, Clone: SYK-01, Abnova™

Catalog No. 89108276 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-276 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-276 Supplier Abnova Corporation Supplier No. MAB0948
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant SYK.

Specifications

Antigen SYK
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone SYK-01
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant SYK.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene SYK
Gene Alias DKFZp313N1010/FLJ25043/FLJ37489
Gene Symbols SYK
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 5-360 of human SYK.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6850
Test Specificity This antibody reacts with protein tyrosine kinase p72Syk (Syk; Syk family tyrosine-specific phospho-transferase), which is required for the transduction of signals through the B cell antigen receptor (BCR) and the high affinity IgE receptor (Fc epsilon RI).
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.