Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYNE1, Mouse, Clone: 4C3, Abnova™

Catalog No. 89106587 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-587 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-587 Supplier Abnova Corporation Supplier No. H00023345M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SYNE1.

This gene encodes a spectrin repeat containing protein expressed in skeletal and smooth muscle, and peripheral blood lymphocytes, that localizes to the nuclear membrane. Mutations in this gene have been associated with autosomal recessive spinocerebellar ataxia 8, also referred to as autosomal recessive cerebellar ataxia type 1 or recessive ataxia of Beauce. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq

Sequence: LRKIQQSVSEFEDKLAVPIKICSSATETYKVLQEHMDLCQALESLSSAITAFSASARKVVNRDSCVQEAAALQQQYEDILRRAKERQTALENLLAHWQRLEKELSSFLTW

Specifications

Antigen SYNE1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4C3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SYNE1.
Formulation ascites with no preservative
Gene SYNE1
Gene Accession No. NM_182961
Gene Alias 8B/CPG2/DKFZp781J13156/FLJ30878/FLJ41140/KIAA0796/KIAA1262/KIAA1756/MYNE1/SCAR8
Gene Symbols SYNE1
Host Species Mouse
Immunogen SYNE1 (NP_892006,1561 a.a. ∽ 1670 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23345
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.