Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYNJ1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105961 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-961 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-961 Supplier Abnova Corporation Supplier No. H00008867A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant SYNJ1.

Synaptojanin 1 is a phosphoinositide phosphatase that regulates levels of membrane phosphatidylinositol-4,5-bisphosphate. As such, expression of this enzyme may affect synaptic transmission and membrane trafficking.[supplied by RefSeq

Sequence: KISNPKGWVTFEEEEDFGVKGKSKSACSDLLGNQPSSFSGSNLTLNDDWNKGTNVSFCVLPSRRPPPPPVPLLPPGTSPPVDPFTTLASKASPTLDFTER

Specifications

Antigen SYNJ1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant SYNJ1.
Formulation 50% glycerol
Gene SYNJ1
Gene Accession No. NM_003895
Gene Alias INPP5G
Gene Symbols SYNJ1
Host Species Mouse
Immunogen SYNJ1 (NP_003886, 1474 a.a. ∽ 1573 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8867
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.