Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SYP, Mouse, Clone: 7H12, Abnova™

Catalog No. 89108556 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-556 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-556 Supplier Abnova Corporation Supplier No. MAB10369
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant SYP.

Synaptophysin (p38) is an integral membrane protein of small synaptic vesicles in brain and endocrine cells.[supplied by OMIM

Specifications

Antigen SYP
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 7H12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant SYP.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene SYP
Gene Symbols SYP
Host Species Mouse
Immunogen Recombinant protein corresponding to human SYP.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6855
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.