Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tacstd1, Mouse, Clone: SPM534, Abnova™

Catalog No. 89110220 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-220 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-220 Supplier Abnova Corporation Supplier No. MAB12078
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Tacstd1.

Specifications

Antigen Tacstd1
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone SPM534
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Tacstd1.
Dilution Immunohistochemistry (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Bioreactor Concentrate with 0.05% azide
Gene Tacstd1
Gene Alias Egp314
Gene Symbols Tacstd1
Host Species Mouse
Immunogen Rat small intestine cells.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171577
Target Species Rat
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.