Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TADA2L, Mouse, Clone: S1, Abnova™

Catalog No. 89105548 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-548 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-548 Supplier Abnova Corporation Supplier No. H00006871M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant TADA2L.

Many DNA-binding transcriptional activator proteins enhance the initiation rate of RNA polymerase II-mediated gene transcription by interacting functionally with the general transcription machinery bound at the basal promoter. Adaptor proteins are usually required for this activation, possibly to acetylate and destabilize nucleosomes, thereby relieving chromatin constraints at the promoter. The protein encoded by this gene is a transcriptional activator adaptor and has been found to be part of the PCAF histone acetylase complex. Two transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq

Sequence: MDRLGSFSNDPSDKPPCRGCSSYLMEPYIKCAECGPPPFFLCLQCFTRGFEYKKHRSDHTYEIMTSDFPVLDPSWTAQEEMALLEAVMDCGFGNWQDVANQMCTKTKEECEKHYMKYFINNPLFASTLLNLKQAEEAKTADTAIPFHSTDDPPRPTFDSLLSRDMAGYMPARADFIEEFDNYAEWDLRDIDFVEDDSDILHALKMAVVDIYHSRLKERQRRKKIIRDHGLINLRKFQLMERRYPKEVQDLYETMRRFARIVGPVEHDKFIESHACRWFLSLEQYLCVYIYINRRDNGVFYVKFYK

Specifications

Antigen TADA2L
Applications ELISA, Western Blot
Classification Monoclonal
Clone S1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant TADA2L.
Formulation ascites with no preservative
Gene TADA2L
Gene Accession No. BC001172
Gene Alias ADA2/ADA2A/FLJ12705/KL04P/hADA2
Gene Symbols TADA2L
Host Species Mouse
Immunogen TADA2L (AAH01172,1 a.a. ∽ 305 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6871
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.