Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAF5L, Mouse, Clone: 1C5, Abnova™

Catalog No. 89106741 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-741 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-741 Supplier Abnova Corporation Supplier No. H00027097M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant TAF5L.

The product of this gene belongs to the WD-repeat TAF5 family of proteins. This gene encodes a protein that is a component of the PCAF histone acetylase complex. The PCAF histone acetylase complex, which is composed of more than 20 polypeptides some of which are TAFs, is required for myogenic transcription and differentiation. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors to facilitate complex assembly and transcription initiation. The encoded protein is structurally similar to one of the histone-like TAFs, TAF5. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq

Sequence: MKRVRTEQIQMAVSCYLKRRQYVDSDGPLKQGLRLSQTAEEMAANLTVQSESGCANIVSAAPCQAEPQQYEVQFGRLRNFLTDSDSQHSHEVMPLLYPLFVYLHLNLVQNSPKSTVE

Specifications

Antigen TAF5L
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1C5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant TAF5L.
Formulation ascites with no preservative
Gene TAF5L
Gene Accession No. NM_014409
Gene Alias PAF65B
Gene Symbols TAF5L
Host Species Mouse
Immunogen TAF5L (NP_055224,1 a.a. ∽ 117 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27097
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.