Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAG-72, Mouse, Clone: B72.3, Abnova™

Catalog No. 89134853 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-853 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-853 Supplier Abnova Corporation Supplier No. MAB11289
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against TAG-72.

Specifications

Antigen TAG-72
Applications Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone B72.3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against TAG-72.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:400) The optimal working dilution should be determined by the end user.
Formulation In 0.2mg/mL PBS (0.05% BSA, 0.05% sodium azide)
Host Species Mouse
Immunogen Membrane enriched fraction of human breast carcinoma liver metastasis
Purification Method Protein G affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.