Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAZ, Mouse, Clone: 4A10-H3, Abnova™

Catalog No. 89105555 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-555 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-555 Supplier Abnova Corporation Supplier No. H00006901M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant TAZ.

This gene encodes a protein that is expressed at high levels in cardiac and skeletal muscle. Mutations in this gene have been associated with a number of clinical disorders including Barth syndrome, dilated cardiomyopathy (DCM), hypertrophic DCM, endocardial fibroelastosis, and left ventricular noncompaction (LVNC). Multiple transcript variants encoding different isoforms have been described. A long form and a short form of each of these isoforms is produced; the short form lacks a hydrophobic leader sequence and may exist as a cytoplasmic protein rather than being membrane-bound. Other alternatively spliced transcripts have been described but the full-length nature of all these transcripts is not known. [provided by RefSeq

Sequence: MPLHVKWPFPAVPPLTWTLASSVVMGLVGTYSCFWTSEWAQAEAGPPGYPCPAGGILKLRHIWNLKLMRWTPAAADICFTKELHSHFFSLGKCVPVCRGDGVYQKGMDFILEKLNHGDWVHIFPEGIGRLIAECHLNPIILPLWHVGEPGDGDREMASGVGGLGLPLVPGCPAPPHVWPSVHCAAGMNDVLPNSPPYFPRFGQKITVLIGKPFSALPVLERLRAENKSAVEMRKALTDFIQEEFQHLKTQAEQLHNHLQPGR

Specifications

Antigen TAZ
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4A10-H3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant TAZ.
Formulation ascites with no preservative
Gene TAZ
Gene Accession No. BC011515
Gene Alias BTHS/CMD3A/EFE/EFE2/FLJ27390/G4.5/LVNCX/Taz1/XAP-2
Gene Symbols TAZ
Host Species Mouse
Immunogen TAZ (AAH11515,1 a.a. ∽ 262 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6901
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.