Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBL1XR1, Mouse, Clone: 3G7, Abnova™

Catalog No. 89107335 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-335 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-335 Supplier Abnova Corporation Supplier No. H00079718M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant TBL1XR1.

The protein encoded by this gene has sequence similarity with members of the WD40 repeat-containing protein family. The WD40 group is a large family of proteins, which appear to have a regulatory function. It is believed that the WD40 repeats mediate protein-protein interactions and members of the family are involved in signal transduction, RNA processing, gene regulation, vesicular trafficking, cytoskeletal assembly and may play a role in the control of cytotypic differentiation. [provided by RefSeq

Sequence: LSLIDAVMPDVVQTRQQAYRDKLAQQQAAAAAAAAAAASQQGSAKNGENTANGEENGAHTIANNHTDMMEVDGDVEIPPNKAVVLRGHESEVFICAWN

Specifications

Antigen TBL1XR1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3G7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant TBL1XR1.
Formulation ascites with no preservative
Gene TBL1XR1
Gene Accession No. NM_024665
Gene Alias C21/DC42/FLJ12894/IRA1/TBLR1
Gene Symbols TBL1XR1
Host Species Mouse
Immunogen TBL1XR1 (NP_071347,81 a.a. ∽ 178 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79718
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.