Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCAP, Mouse, Clone: 1A3, Abnova™

Catalog No. 89105888 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-888 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-888 Supplier Abnova Corporation Supplier No. H00008557M13A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant TCAP.

Sarcomere assembly is regulated by the muscle protein titin. Titin is a giant elastic protein with kinase activity that extends half the length of a sarcomere. It serves as a scaffold to which myofibrils and other muscle related proteins are attached. This gene encodes a protein found in striated and cardiac muscle that binds to the titin Z1-Z2 domains and is a substrate of titin kinase, interactions thought to be critical to sarcomere assembly. Mutations in this gene are associated with limb-girdle muscular dystrophy type 2G. [provided by RefSeq

Sequence: MATSELSCEVSEENCERREAFWAEWKDLTLSTRPEEGCSLHEEDTQRHETYHQQGQCQVLVQRSPWLMMRMGILGRGLQEYQLPYQRVLPLPIFTPAKMG

Specifications

Antigen TCAP
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1A3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant TCAP.
Formulation ascites with no preservative
Gene TCAP
Gene Accession No. NM_003673
Gene Alias CMD1N/LGMD2G/T-cap/TELE/telethonin
Gene Symbols TCAP
Host Species Mouse
Immunogen TCAP (NP_003664.1,1 a.a. ∽ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8557
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.