Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-TCL1A, Clone: TCL1A, Alexa Fluor 488,

Catalog No. MABF1214AF4 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 488
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABF1214AF4 100 μL Alexa Fluor 488
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF1214AF4 Supplier MilliporeSigma™ Supplier No. MABF1214AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects TCL1A clone: TCL1A in Human samples, and it is validated for Immunocytochemistry

T-cell leukemia/lymphoma protein 1A (UniProt P56279; also known as Oncogene TCL-1, Oncogene TCL1, Protein p14 TCL1, T-cell lymphoma-1) is encoded by the TCL1A (also known as TCL1) gene (Gene ID 8115) in human. TCL1A belongs to the T-cell leukemia/lymphoma 1 (TCL1) family of oncoproteins that enhance cell proliferation and survival by augmenting AKT signal transduction. TCL1A and the related TCL1B play key roles in several B and T cell malignancies. TCL1A does not exhibit enzymatic activity and functions as an AKT activity enhancer via binding to its pleckstrin homology (PH) domain, resulting in enhanced phosphorylation of downstream AKT substrates, such as glycogen synthase kinase-3 beta (GSK3beta). TCL1A is also reported to interact with IkappaB, albeit as a ∼100-fold lower affinity than NF-kappaB.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCL1A
Applications Immunocytochemistry
Classification Monoclonal
Clone TCL1A
Conjugate Alexa Fluor 488
Formulation Purified mouse antibody conjugate in PBS with 15mg/mL BSA and 0.1% sodium azide.
Gene Accession No. P56279
Gene Symbols TCL1A; TCL1
Host Species Mouse
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 60-73 (RPMTPTQIGPSLLP) of human TCL1A.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NM_021966
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.