Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR alpha/beta, Mouse, Clone: KJ12-98, Abnova™

Catalog No. 89135197 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-197 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-197 Supplier Abnova Corporation Supplier No. MAB2457
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against TCR alpha/beta.

Specifications

Antigen TCR alpha/beta
Applications ELISA, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone KJ12-98
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against TCR alpha/beta.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Host Species Mouse
Immunogen Native from a mouse T cell hybridoma 3DT-52.5.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The 3DT-52.5 TcR idiotype in mouse.
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.