Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-TDRD3, Clone: 2E11,

Catalog No. MABE1042MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1042MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1042MI Supplier MilliporeSigma™ Supplier No. MABE1042
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects TDRD3 clone: 2E11 in Human samples, and it is validated for Western Blotting

Tudor domain-containing protein 3 (UniProt Q9H7E2) is encoded by the TDRD3 gene in human (Entrez Gene ID 81550). TDRD3 belongs to an evolutionarily conserved family of 12 proteins (TDRD1-TDRD12) that contain one or more Tudor domains, which recognize symmetric dimethylarginine (sDMA) residues with the exception of TDRD3. Unlike other TDRD members, TDRD3 associates selectively with asymmetric dimethylarginine (aDMA or Rme2a) on histones and on RNA Polymerase II (RNAP II) C-terminal domain (CTD), but not with other methylarginine motifs. TDRD3 is reported to act as a positive regulator of c-MYC gene transcription by recruiting topoisomerase IIIB (TOP3B) to the c-MYC gene promoter region via H4R3me2a recognition. (PMID 23066109 & 24507716). Anti-TDRD3, clone 2E11, is a purified mouse monoclonal antibody (PMID 21172665) against an epitope within a region in the C-terminal half of all three human TDRD3 spliced isoforms (UniProt Q9H7E2). Clone 2E11 is suitable for Western blotting application (PMID 21172665 & 24507716).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDRD3
Applications Western Blot
Classification Monoclonal
Clone 2E11
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q9H7E2
Gene Symbols TDRD3
Host Species Mouse
Immunogen Recombinant protein corresponding to human TDRD3.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001139542
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.