Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TEC, Mouse, Clone: 3A5, Abnova™

Catalog No. 89108558 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-558 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-558 Supplier Abnova Corporation Supplier No. MAB10371
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant TEC.

The protein encoded by this gene belongs to the Tec family of nonreceptor protein-tyrosine kinases containing a pleckstrin homology domain. Tec family kinases are involved in the intracellular signaling mechanisms of cytokine receptors, lymphocyte surface antigens, heterotrimeric G-protein coupled receptors, and integrin molecules. They are also key players in the regulation of the immune functions. Tec kinase is an integral component of T cell signaling and has a distinct role in T cell activation. This gene may be associated with myelodysplastic syndrome. [provided by RefSeq

Specifications

Antigen TEC
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3A5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant TEC.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene TEC
Gene Alias MGC126760/MGC126762/PSCTK4
Gene Symbols TEC
Host Species Mouse
Immunogen Recombinant protein corresponding to human TEC.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7006
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.