Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TECTA, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105572 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-572 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-572 Supplier Abnova Corporation Supplier No. H00007007A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant TECTA.

The tectorial membrane is an extracellular matrix of the inner ear that contacts the stereocilia bundles of specialized sensory hair cells. Sound induces movement of these hair cells relative to the tectorial membrane, deflects the stereocilia, and leads to fluctuations in hair-cell membrane potential, transducing sound into electrical signals. Alpha-tectorin is one of the major noncollagenous components of the tectorial membrane. Mutations in the TECTA gene have been shown to be responsible for autosomal dominant nonsyndromic hearing impairment and a recessive form of sensorineural pre-lingual nonsyndromic deafness. [provided by RefSeq

Sequence: YATPTRDSNDKLRYFIIEGGCQNLKDNTIGIEENAVSLTCRFHVTVFKFIGDYDEVHLHCAVSLCDSEKYSCKITCPHNSRIATDYTKEPKEQIISVGPI

Specifications

Antigen TECTA
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant TECTA.
Formulation 50% glycerol
Gene TECTA
Gene Accession No. NM_005422
Gene Alias DFNA12/DFNA8/DFNB21
Gene Symbols TECTA
Host Species Mouse
Immunogen TECTA (NP_005413, 1981 a.a. ∽ 2080 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7007
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.