Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TERF1, Mouse, Clone: 3H11, Abnova™

Catalog No. 89111024 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-024 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-024 Supplier Abnova Corporation Supplier No. MAB2481
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against TERF1.

This gene encodes a telomere specific protein which is a component of the telomere nucleoprotein complex. This protein is present at telomeres throughout the cell cycle and functions as an inhibitor of telomerase, acting in cis to limit the elongation of individual chromosome ends. The protein structure contains a C-terminal Myb motif, a dimerization domain near its N-terminus and an acidic N-terminus. Two transcripts of this gene are alternatively spliced products. [provided by RefSeq

Specifications

Antigen TERF1
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3H11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against TERF1.
Dilution Western Blot (1:100-1:200) Immunofluorescence (1:100-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene TERF1
Gene Alias FLJ41416/PIN2/TRBF1/TRF/TRF1/hTRF1-AS/t-TRF1
Gene Symbols TERF1
Host Species Mouse
Immunogen Human TERF1.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7013
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.