Learn More
Terminal-Deoxynucleotidyl Transferase Mouse anti-Human, APC, Clone: E17-1519, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 347196
Description
Anti-TdT, clone E17-1519, is generated from the fusion of FO myeloma cells with spleen cells from BALB/c mice immunized with purified TdT enzyme.
Anti–terminal-deoxynucleotidyl transferase (TdT) recognizes a 60-kilodalton (kd) polymerase, a nuclear enzyme that catalyzes the template-independent addition of nucleotides to single-stranded DNA primers. It has been reported that TdT is involved in the regulation or translocation or both of DNA and gene rearrangement during normal T- and B-cell development.

Specifications
Terminal-Deoxynucleotidyl Transferase | |
Monoclonal | |
25μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
Purified Human TdT | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry, Immunocytochemistry | |
E17-1519 | |
APC | |
Mouse | |
100 Tests | |
Primary | |
Store at 2-8°C. Protect from light. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.