Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TES, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106684 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-684 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-684 Supplier Abnova Corporation Supplier No. H00026136A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant TES.

Cancer-associated chromosomal changes often involve regions containing fragile sites. This gene maps to a commom fragile site on chromosome 7q31.2 designated FRA7G. This gene is similar to mouse Testin, a testosterone-responsive gene encoding a Sertoli cell secretory protein containing three LIM domains. LIM domains are double zinc-finger motifs that mediate protein-protein interactions between transcription factors, cytoskeletal proteins and signaling proteins. Multiple protein isoforms are encoded by transcript variants of this gene. [provided by RefSeq

Sequence: MDLENKVKKMGLGHEQGFGAPCLKCKEKCEGFELHFWRKICRNCKCGQEEHDVLLSNEEDRKVGKLFEDTKYTTLIAKLKSDGIPMYKRNVMILTNPVAAKKNVSINTVTYEWAPPVQNQALARQYMQMLPKEKQPVAGSEGAQYRKKQLAKQLPAHDQDPSKCHELSPREVKEMEQFVKKYKSEALGVGDVKLPCEMDAQGPKQMNIPGGDRSTPAAVGAMEDKSAEHKRTQYSCYCCKLSMKEGDPAIYAERAGYDKLWHPACFVCSTCHELLVDMIYFWKNEKLYCGRHYCDSEKPRCAGCDELIFSNEYTQAENQNWHLKHFCCFDCDSILAGEIYVMVNDKPVCKPCYVKNHAVVCQGCHNAIDPEVQRVTYNNFSWHASTECFLCSCCSKCLIGQKFMPVEGMVFCSVECKKRMS

Specifications

Antigen TES
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant TES.
Formulation 50% glycerol
Gene TES
Gene Accession No. BC001451
Gene Alias DKFZp586B2022/MGC1146/TESS/TESS-2
Gene Symbols TES
Host Species Mouse
Immunogen TES (AAH01451, 1 a.a. ∽ 421 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26136
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.