Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TF, Mouse, Clone: PTF-02, Abnova™

Catalog No. 89111511 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-511 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-511 Supplier Abnova Corporation Supplier No. MAB3642
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native TF.

Specifications

Antigen TF
Applications ELISA, Western Blot
Classification Monoclonal
Clone PTF-02
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native TF.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene TF
Gene Symbols TF
Host Species Mouse
Immunogen Native purified TF from porcine transferrin.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 396996
Test Specificity This antibody recognizes porcine serum transferrin, a 77 KDa single polypeptide chain glycoprotein (member of the iron binding family of proteins). It is synthesised in the liver and consists of two domains each having a high affinity reversible binding site for Fe3+.
Target Species Pig
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.