Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TF, Mouse, Monoclonal Antibody, Abnova™

Catalog No. 89113399 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-399 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-399 Supplier Abnova Corporation Supplier No. MAB8253
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native TF.

This gene encodes a glycoprotein with an approximate molecular weight of 76.5kDa. It is thought to have been created as a result of an ancient gene duplication event that led to generation of homologous C and N-terminal domains each of which binds one ion of ferric iron. The function of this protein is to transport iron from the intestine, reticuloendothelial system, and liver parenchymal cells to all proliferating cells in the body. This protein may also have a physiologic role as granulocyte/pollen-binding protein (GPBP) involved in the removal of certain organic matter and allergens from serum. [provided by RefSeq

Specifications

Antigen TF
Applications ELISA, Immunofluorescence
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native TF.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene TF
Gene Alias DKFZp781D0156/PRO1557/PRO2086
Gene Symbols TF
Host Species Mouse
Immunogen Native purified human TF.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7018
Test Specificity This antibody reacts with the N-terminal domain of human transferrin. This antibody does not cross-react with mouse, rabbit, or bovine transferrin, nor with human lactoferrin.
Target Species Human
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.