Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFAP2A, Mouse, Clone: AP2a 8G8/5, Abnova™

Catalog No. 89136331 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-331 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-331 Supplier Abnova Corporation Supplier No. MAB7292
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant TFAP2A.

Specifications

Antigen TFAP2A
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone AP2a 8G8/5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant TFAP2A.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS
Host Species Mouse
Immunogen Recombinant fusion protein corresponding to bacterial TFAP2A.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Does not cross-react with AP2 beta or Ap2 gamma. Not yet tested in other species. Predicted molecular weight: 50 KDa.
Target Species Human
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.