Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFAP4, Mouse, Clone: 7C5, Abnova™

Catalog No. 89105576 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-576 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-576 Supplier Abnova Corporation Supplier No. H00007023M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant TFAP4.

Transcription factors of the basic helix-loop-helix-zipper (bHLH-ZIP) family contain a basic domain, which is used for DNA binding, and HLH and ZIP domains, which are used for oligomerization. Transcription factor AP4 activates both viral and cellular genes by binding to the symmetrical DNA sequence CAGCTG (Mermod et al., 1988 [PubMed 2833704]; Hu et al., 1990 [PubMed 2123466]).[supplied by OMIM

Sequence: AEYIFSLEQEKTRLLQQNTQLKRFIQELSGSSPKRRRAEDKDEGIGSPDIWEDEKAEDLRREMIELRQQLDKERSVRMMLEEQVRSLEAHMYPEKLKVIA

Specifications

Antigen TFAP4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 7C5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant TFAP4.
Formulation ascites with no preservative
Gene TFAP4
Gene Accession No. NM_003223
Gene Alias AP-4/bHLHc41
Gene Symbols TFAP4
Host Species Mouse
Immunogen TFAP4 (NP_003214,93 a.a. ∽ 192 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7023
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.