Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFE3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105579 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-579 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-579 Supplier Abnova Corporation Supplier No. H00007030A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant TFE3.

TFE3, a member of the helix-loop-helix family of transcription factors, binds to the mu-E3 motif of the immunoglobulin heavy-chain enhancer and is expressed in many cell types (Henthorn et al., 1991 [PubMed 1685140]).[supplied by OMIM

Sequence: RPPPAQVPREVLKVQTHLENPTRYHLQQARRQQVKQYLSTTLGPKLASQALTPPPGPASAQPLPAPEAAHTTGPTGSAPNSPMALLTIG

Specifications

Antigen TFE3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant TFE3.
Formulation 50% glycerol
Gene TFE3
Gene Accession No. NM_006521
Gene Alias RCCP2/TFEA/bHLHe33
Gene Symbols TFE3
Host Species Mouse
Immunogen TFE3 (NP_006512, 166 a.a. ∽ 254 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7030
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.