Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFF1, Mouse, Clone: GE2, Abnova™

Catalog No. 89109514 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-514 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-514 Supplier Abnova Corporation Supplier No. MAB11350
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of TFF1.

Members of the trefoil family are characterized by having at least one copy of the trefoil motif, a 40-amino acid domain that contains three conserved disulfides. They are stable secretory proteins expressed in gastrointestinal mucosa. Their functions are not defined, but they may protect the mucosa from insults, stabilize the mucus layer, and affect healing of the epithelium. This gene, which is expressed in the gastric mucosa, has also been studied because of its expression in human tumors. This gene and two other related trefoil family member genes are found in a cluster on chromosome 21. [provided by RefSeq

Sequence: CFDDTVRGVPWCFYPNTIDVPPEEECEF

Specifications

Antigen TFF1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone GE2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of TFF1.
Dilution Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (0.5-1 ug/mL for 30 min at RT) Immunoprecipitation (0.5-1 ug/500 ug protein lysate) The optimal working dilution should be determined by the end user
Formulation In PBS, pH 7.4 (0.05% BSA, 0.05% sodium azide)
Gene TFF1
Gene Alias BCEI/D21S21/HP1.A/HPS2/pNR-2/pS2
Gene Symbols TFF1
Host Species Mouse
Immunogen A Synthetic peptide corresponding to amino acids 57-84 at C-terminus of human TFF1.
Purification Method Protein A/G affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7031
Target Species Human
Content And Storage Store at 4°C.
Avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.