Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFF1, Mouse, Clone: M2, Abnova™

Catalog No. 89105581 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-581 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-581 Supplier Abnova Corporation Supplier No. H00007031M04A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant TFF1.

Members of the trefoil family are characterized by having at least one copy of the trefoil motif, a 40-amino acid domain that contains three conserved disulfides. They are stable secretory proteins expressed in gastrointestinal mucosa. Their functions are not defined, but they may protect the mucosa from insults, stabilize the mucus layer, and affect healing of the epithelium. This gene, which is expressed in the gastric mucosa, has also been studied because of its expression in human tumors. This gene and two other related trefoil family member genes are found in a cluster on chromosome 21. [provided by RefSeq

Sequence: MATMENKVICALVLVSMLALGTLAEAQTETCTVAPRERQNCGFPGVTPSQCANKGCCFDDTVRGVPWCFYPNTIDVPPEEECEF

Specifications

Antigen TFF1
Applications ELISA, Western Blot
Classification Monoclonal
Clone M2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant TFF1.
Formulation ascites with no preservative
Gene TFF1
Gene Accession No. BC032811
Gene Alias BCEI/D21S21/HP1.A/HPS2/pNR-2/pS2
Gene Symbols TFF1
Host Species Mouse
Immunogen TFF1 (AAH32811.1,1 a.a. ∽ 84 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7031
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.