Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFRC, Mouse, Biotin, Clone: RVS10, Abnova™

Catalog No. 89112683 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-683 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-683 Supplier Abnova Corporation Supplier No. MAB6077
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native TFRC.

Specifications

Antigen TFRC
Applications Flow Cytometry
Classification Monoclonal
Clone RVS10
Conjugate Biotin
Description Mouse monoclonal antibody raised against native TFRC.
Dilution Flow Cytometry (10 ul/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene TFRC
Gene Alias CD71/TFR/TFR1/TRFR
Gene Symbols TFRC
Host Species Mouse
Immunogen Native purified from NALM-6 cells.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7037
Test Specificity Specificity human CD71
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.