Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TG, Mouse, Clone: 5E6, Abnova™

Catalog No. 89108017 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-017 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-017 Supplier Abnova Corporation Supplier No. MAB0637

Mouse monoclonal antibody raised against native TG.

Specifications

Antigen TG
Applications ELISA
Classification Monoclonal
Clone 5.00E+06
Conjugate Unconjugated
Description thyroglobulin
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene TG
Gene Alias AITD3, TGN
Gene Symbols TG
Host Species Mouse
Immunogen Native purified human TG.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7038
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.