Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-TGF-alpha, Clone: TG86+P/T1, Azide Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Specifications

Antigen TGF-alpha
Clone TG86+P/T1
Concentration 1.0 mg/mL
Dilution Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 1-2ug/500ug protein, Immunohistochemistry-Paraffin 1-2ug/ml, Immunohistochemistry-Frozen 1-2ug/ml, CyTOF-ready
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP23472302
SDP
View Documents
Novus Biologicals
NBP2347230.2 MG
0.2 mg N/A N/A N/A
Description

Description

TGF-alpha Monoclonal antibody specifically detects TGF-alpha in Human, Rabbit, Zebrafish samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.
Specifications

Specifications

TGF-alpha
1.0 mg/mL
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Angiogenesis, Cell Cycle and Replication, Signal Transduction
10mM PBS - BSA free with No Preservative
protransforming growth factor alpha, TFGA, transforming growth factor, alpha, transforming growth factor-alpha
TGFA
IgM
Protein A purified
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
6 kDa
TG86+P/T1
Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 1-2ug/500ug protein, Immunohistochemistry-Paraffin 1-2ug/ml, Immunohistochemistry-Frozen 1-2ug/ml, CyTOF-ready
Monoclonal
Purified
RUO
Human, Rabbit, Zebrafish
P01135
7039
A 10-amino acid synthetic peptide (aa34-43) from human TGFalpha (TG86 and P/T1)
Primary
Protein A purified
This antibody reacts with the TGF alpha and shows no cross-reaction with EGF and the neuropeptide synenkephalin. The staining with this MAb is completely blocked by the peptide used for raising this antibody.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.