Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGFa, Mouse, Clone: MF9, Abnova™

Catalog No. 89109509 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-509 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-509 Supplier Abnova Corporation Supplier No. MAB11345
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of Tgfa.

Specifications

Antigen TGFa
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MF9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of Tgfa.
Dilution Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Frozen sections) (1-2 ug/mL for 30 min at RT) Immunoprecipitation (1-2 ug/500 ug protein) Western Blot (0.25-0.5 ug/mL) The optimal working dilutio
Formulation In PBS, pH 7.4 (0.05% BSA, 0.05% sodium azide)
Gene Tgfa
Gene Alias RATTGFAA/TGFAA
Gene Symbols Tgfa
Host Species Mouse
Immunogen A synthetic peptide corresponding to C-terminus of rat Tgfa.
Purification Method Protein A/G affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24827
Target Species Bovine, Human, Mouse, Rat
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.