Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGFB1, Mouse, Clone: 2C6, Abnova™

Catalog No. 89112821 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-821 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-821 Supplier Abnova Corporation Supplier No. MAB6319
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native TGFB1.

TGFB is a multifunctional peptide that controls proliferation, differentiation, and other functions in many cell types. TGFB acts synergistically with TGFA (MIM 190170) in inducing transformation. It also acts as a negative autocrine growth factor. Dysregulation of TGFB activation and signaling may result in apoptosis. Many cells synthesize TGFB and almost all of them have specific receptors for this peptide. TGFB1, TGFB2 (MIM 190220), and TGFB3 (MIM 190230) all function through the same receptor signaling systems.[supplied by OMIM

Specifications

Antigen TGFB1
Applications ELISA, Immunohistochemistry, Inhibition Assays, Western Blot
Classification Monoclonal
Clone 2C6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native TGFB1.
Dilution Western Blot (50-500 ng/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS, pH 7.0
Gene TGFB1
Gene Alias CED/DPD1/TGFB/TGFbeta
Gene Symbols TGFB1
Host Species Mouse
Immunogen Native purified human platelets TGFB1.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7040
Test Specificity This antibody reacts with the dimeric (25 KDa) and monomeric (12.5 KDa) forms of TGFB1 under both non-reducing and reducing conditions respectively.
Target Species Human
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.