Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGFB1, Mouse, Clone: 3F12, Abnova™

Catalog No. 89105588 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-588 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-588 Supplier Abnova Corporation Supplier No. H00007040M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant TGFB1.

TGFB is a multifunctional peptide that controls proliferation, differentiation, and other functions in many cell types. TGFB acts synergistically with TGFA (MIM 190170) in inducing transformation. It also acts as a negative autocrine growth factor. Dysregulation of TGFB activation and signaling may result in apoptosis. Many cells synthesize TGFB and almost all of them have specific receptors for this peptide. TGFB1, TGFB2 (MIM 190220), and TGFB3 (MIM 190230) all function through the same receptor signaling systems.[supplied by OMIM

Sequence: ALDTNYCFSSTEKNCCVRQLYIDFRKDLGWKWIHEPKGYHANFCLGPCPYIWSLDTQYSKVLALYNQHNPGASAAPCCVPQALEPLPIVYYVGRKPKVEQLSNMIVRSCKCS

Specifications

Antigen TGFB1
Applications ELISA
Classification Monoclonal
Clone 3F12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant TGFB1.
Formulation ascites with no preservative
Gene TGFB1
Gene Accession No. NM_000660
Gene Alias CED/DPD1/TGFB/TGFbeta
Gene Symbols TGFB1
Host Species Mouse
Immunogen TGFB1 (NP_000651,279 a.a. ∽ 390 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7040
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.