Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGFB1, Mouse, Clone: 7E9, Abnova™

Catalog No. 89110354 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-354 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-354 Supplier Abnova Corporation Supplier No. MAB1317
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant TGFB1.

TGFB is a multifunctional peptide that controls proliferation, differentiation, and other functions in many cell types. TGFB acts synergistically with TGFA (MIM 190170) in inducing transformation. It also acts as a negative autocrine growth factor. Dysregulation of TGFB activation and signaling may result in apoptosis. Many cells synthesize TGFB and almost all of them have specific receptors for this peptide. TGFB1, TGFB2 (MIM 190220), and TGFB3 (MIM 190230) all function through the same receptor signaling systems.[supplied by OMIM

Specifications

Antigen TGFB1
Applications ELISA
Classification Monoclonal
Clone 7E9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant TGFB1.
Dilution ELISA (1:10000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene TGFB1
Gene Alias CED/DPD1/TGFB/TGFbeta
Gene Symbols TGFB1
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human TGFB1.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7040
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.