Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGIF1, Mouse, Clone: 1D12, Abnova™

Catalog No. 89105590 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-590 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-590 Supplier Abnova Corporation Supplier No. H00007050M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant TGIF1.

The protein encoded by this gene is a member of the three-amino acid loop extension (TALE) superclass of atypical homeodomains. TALE homeobox proteins are highly conserved transcription regulators. This particular homeodomain binds to a previously characterized retinoid X receptor responsive element from the cellular retinol-binding protein II promoter. In addition to its role in inhibiting 9-cis-retinoic acid-dependent RXR alpha transcription activation of the retinoic acid responsive element, the protein is an active transcriptional co-repressor of SMAD2 and may participate in the transmission of nuclear signals during development and in the adult. Mutations in this gene are associated with holoprosencephaly type 4, which is a structural anomaly of the brain. Alternative splicing has been observed at this locus and eight variants, encoding four distinct isoforms, are described. [provided by RefSeq

Sequence: PGSVLARPSVICHTTVTALKDVPFSLCQSVGVGQNTDIQQIAAKNFTDTSLMYPEDTCKSGPSTNTQSGLFNTPPPTPPDLNQDFSGFQLLVDVALKRAAEMELQAKLTA

Specifications

Antigen TGIF1
Applications ELISA
Classification Monoclonal
Clone 1D12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant TGIF1.
Formulation ascites with no preservative
Gene TGIF1
Gene Accession No. NM_003244
Gene Alias HPE4/MGC39747/MGC5066/TGIF
Gene Symbols TGIF1
Host Species Mouse
Immunogen TGIF1 (NP_003235,163 a.a. ∽ 272 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7050
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.