Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGM2, Mouse, Clone: XTG5, Abnova™

Catalog No. 89135339 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-339 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-339 Supplier Abnova Corporation Supplier No. MAB3046

Mouse monoclonal antibody raised against native TGM2.

Specifications

Antigen TGM2
Applications Immunohistochemistry
Classification Monoclonal
Clone XTG5
Conjugate Unconjugated
Formulation In 10mM Tris, 150mM NaCl, pH 7.5 (0.05% sodium azide)
Host Species Mouse
Immunogen Native purified TGM2.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.