Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Th, Mouse, Clone: 1B5, Abnova™

Catalog No. 89113702 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
3 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-702 3 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-702 Supplier Abnova Corporation Supplier No. MAB9597

Mouse monoclonal antibody raised against partial recombinant Th.

Specifications

Antigen Th
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1B5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant Th.
Dilution Immunohistochemistry (1:20-1:40) Western Blot (1:25-1:50) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from tissue culture supernatant (0.09% sodium azide)
Gene Th
Gene Symbols Th
Host Species Mouse
Immunogen Recombinant protein corresponding to a portion at C-terminus of mouse Th.
Quantity 3 x 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21823
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with sterile distilled water store at 4°C for at least two months. For long term storage store at -20°C or below. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.