Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-THOP1, Clone: 4D6, Abnova™

Catalog No. 89110908 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-908 Supplier Abnova Corporation Supplier No. MAB2290
Add to Cart
Add to Cart

Specifications

Classification Monoclonal
Clone 4D6
Description Mouse monoclonal antibody raised against synthetic peptide of THOP1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:400) Western Blot (1:750-1:3000) The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine, 150mM NaCl (0.05% sodium azide)
Gene Alias EP24.15/MEPD_HUMAN/MP78/TOP
Gene Symbols THOP1
Immunogen A synthetic peptide corresponding to amino acids 56-72 of human THOP1.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 7064
Test Specificity This antibody is specific to thimet oligopeptidase.
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.