Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THY1, Mouse, Clone: aTHy-1A1, Abnova™

Catalog No. 89113158 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-158 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-158 Supplier Abnova Corporation Supplier No. MAB7281

Mouse monoclonal antibody raised against full-length native THY1.

Specifications

Antigen THY1
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone aTHy-1A1
Conjugate Unconjugated
Description Thy-1 cell surface antigen
Formulation In PBS
Gene THY1
Gene Alias CD90, FLJ33325
Gene Symbols THY1
Host Species Mouse
Immunogen Native purified human THY1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7070
Target Species Human
Content And Storage Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.