Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THY1, Mouse, Clone: F15-42-1, Abnova™

Catalog No. 89111054 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-054 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-054 Supplier Abnova Corporation Supplier No. MAB2525
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native THY1.

Specifications

Antigen THY1
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone F15-42-1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native THY1.
Dilution Flow cytometry (10ul of the suggested working dilution 1:10-1:50 to label 106 cells in 100ul) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene THY1
Gene Alias CD90/FLJ33325
Gene Symbols THY1
Host Species Mouse
Immunogen Native purified human THY1.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7070
Test Specificity This antibody recognizes the human CD90 cell surface antigen, a 25kD glycoprotein homologous to rat Thy1.
Target Species Human, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.